Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003986.2 | NP_003977.1 | 8, 9 | 1379 | ||
XM_005253159.2 | XP_005253216.1 | 8, 9 | 1627 | |||
XM_005253160.2 | XP_005253217.1 | 8, 9 | 1624 | |||
XM_005253161.2 | XP_005253218.1 | 8, 9 | 1376 | |||
XM_011520402.1 | XP_011518704.1 | 9, 10 | 1787 | |||
XM_017018403.1 | XP_016873892.1 | 7, 8 | 1309 | |||
GenBank mRNA | AF082868.1 | - | 7, 8 | 1077 | ||
AK290108.1 | - | 7, 8 | 1124 | |||
AK313422.1 | - | 7, 8 | 1127 | |||
BC011034.1 | - | 6, 7 | 1067 | 4042355 | ||
CR456812.1 | - | 6, 7 | 1011 |