Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270965.1 | NP_001257894.1 | 18 | 3280 | ||
NM_001270966.1 | NP_001257895.1 | 18 | 2801 | |||
NM_002779.4 | NP_002770.3 | 17 | 3604 | |||
NR_073110.1 | 8 | 1376 | ||||
XM_011539968.1 | XP_011538270.1 | 18 | 3286 | |||
XM_011539969.2 | XP_011538271.1 | 18 | 3303 | |||
XM_017016433.1 | XP_016871922.1 | 18 | 3297 | |||
XM_017016434.1 | XP_016871923.1 | 17 | 3224 | |||
GenBank mRNA | AB095931.1 | - | 17 | 3604 | ||
BC039259.1 | - | 18 | 2788 | |||
BC142643.1 | - | 18 | 3280 | 40147927 | ||
BC142689.1 | - | 18 | 3280 | 40148656 |