Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270481.1 | NP_001257410.1 | 6 | 736 | ||
NM_002798.2 | NP_002789.1 | 6 | 716 | |||
GenBank mRNA | AK311768.1 | - | 6 | 679 | ||
BC000835.2 | - | 6 | 683 | 3456009 | ||
BM553399.1 | - | 6 | 663 | |||
BQ691763.1 | - | 6 | 698 | |||
BT009809.1 | - | 6 | 665 | |||
BU848746.1 | - | 6 | 668 | |||
CR457065.1 | - | 6 | 665 | |||
D29012.1 | - | 6 | 674 | |||
X61971.1 | - | 4 | 488 |