Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002809.3 | NP_002800.2 | 5 | 1071 | ||
GenBank mRNA | AK022896.1 | - | 5 | 982 | ||
AK054611.1 | - | 5 | 1015 | |||
AK055799.1 | - | 6 | 952 | |||
AK094206.1 | - | 6 | 993 | |||
AK293539.1 | - | 6 | 957 | |||
AK298411.1 | - | 5 | 910 | |||
AK300081.1 | - | 5 | 738 | |||
AK300973.1 | - | 6 | 987 | |||
BC000074.2 | - | 5 | 971 | 3505074 | ||
BC004859.2 | - | 5 | 933 | 3832823 | ||
BC012302.1 | - | 5 | 1019 | 3868681 | ||
BC020518.1 | - | 5 | 969 | 3901555 | ||
BC025686.1 | - | 5 | 969 | 5205360 | ||
BT007217.1 | - | 5 | 868 | |||
CR456772.1 | - | 5 | 868 | |||
D67025.1 | - | 5 | 1025 |