Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002809.3 | NP_002800.2 | 9 | 1443 | ||
GenBank mRNA | AF091075.1 | - | 2 | 151 | ||
AK022896.1 | - | 9 | 1354 | |||
AK054611.1 | - | 9 | 1387 | |||
AK055358.1 | - | 3 | 2215 | |||
AK055799.1 | - | 10 | 1324 | |||
AK094206.1 | - | 10 | 1365 | |||
AK293539.1 | - | 10 | 1329 | |||
AK298411.1 | - | 9 | 1282 | |||
AK300081.1 | - | 9 | 1110 | |||
AK300973.1 | - | 10 | 1359 | |||
BC000074.2 | - | 9 | 1343 | 3505074 | ||
BC004859.2 | - | 9 | 1305 | 3832823 | ||
BC012302.1 | - | 9 | 1391 | 3868681 | ||
BC020518.1 | - | 9 | 1341 | 3901555 | ||
BC025686.1 | - | 9 | 1341 | 5205360 | ||
BT007217.1 | - | 9 | 1240 | |||
CR456772.1 | - | 9 | 1240 | |||
D67025.1 | - | 9 | 1397 |