Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002812.4 | NP_002803.2 | 6 | 891 | ||
GenBank mRNA | AK301771.1 | - | 6 | 902 | ||
AK304486.1 | - | 6 | 476 | |||
AK308809.1 | - | 6 | 980 | |||
BC001164.1 | - | 6 | 642 | 3528096 | ||
BC065006.1 | - | 6 | 630 | 6055235 | ||
BQ684248.1 | - | 6 | 654 | |||
BT019688.1 | - | 6 | 560 | |||
D38047.1 | - | 6 | 630 | |||
EU668354.1 | - | 6 | 891 |