Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261400.2 | NP_001248329.1 | 3 | 416 | ||
NM_002813.6 | NP_002804.2 | 5 | 731 | |||
NR_048555.2 | 4 | 519 | ||||
GenBank mRNA | AK055339.1 | - | 1 | 1687 | ||
AK315389.1 | - | 5 | 700 | |||
BC002383.2 | - | 5 | 691 | 2961610 | ||
BC004184.2 | - | 5 | 691 | 2961610 | ||
BC004213.2 | - | 5 | 691 | 2961610 | ||
BM850893.1 | - | 4 | 440 | |||
BU956605.1 | - | 6 | 713 | |||
CB111716.1 | - | 3 | 361 |