Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316341.1 | NP_001303270.1 | 8 | 918 | ||
NM_002816.4 | NP_002807.1 | 6 | 634 | |||
NM_174871.3 | NP_777360.1 | 5 | 574 | |||
GenBank mRNA | AB003103.1 | - | 6 | 569 | ||
AK091198.1 | - | 5 | 552 | |||
AK222679.1 | - | 6 | 551 | |||
AK307821.1 | - | 6 | 584 | |||
AK308883.1 | - | 6 | 575 | |||
BC019062.2 | - | 6 | 565 | 4111477 | ||
BC065826.1 | - | 8 | 838 | 30386487 | ||
KU178288.1 | - | 6 | 526 | |||
KU178289.1 | - | 6 | 526 |