Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF083245.1 | - | 4 | 308 | ||
AF086708.1 | - | 4 | 254 | |||
AF107837.1 | - | 4 | 287 | |||
AK295944.1 | - | 3 | 293 | |||
AK307447.1 | - | 3 | 221 | |||
BC001100.2 | - | 4 | 309 | 3506530 | ||
BC001747.1 | - | 4 | 277 | 3535807 | ||
BM456586.1 | - | 4 | 327 | |||
BT007307.1 | - | 4 | 254 | |||
BX647314.1 | - | 3 | 231 |