Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002819.4 | NP_002810.1 | 15 | 3148 | ||
NM_031990.3 | NP_114367.1 | 15 | 3127 | |||
NM_031991.3 | NP_114368.1 | 14 | 3070 | |||
XM_005259597.2 | XP_005259654.1 | 15 | 3651 | |||
XM_005259598.2 | XP_005259655.1 | 14 | 3573 | |||
GenBank mRNA | BC002397.2 | - | 15 | 3112 | 2821531 | |
BC004383.1 | - | 14 | 2979 | 3615251 | ||
X60648.1 | - | 14 | 2969 | |||
X62006.1 | - | 14 | 2998 | |||
X65371.1 | - | 15 | 3055 | |||
X65372.1 | - | 15 | 3076 | |||
X66975.1 | - | 15 | 3127 |