Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002841.3 | NP_002832.3 | 7 | 1470 | ||
XM_005265353.3 | XP_005265410.1 | 7 | 1128 | |||
XM_017006961.1 | XP_016862450.1 | 8 | 986 | |||
XM_017006962.1 | XP_016862451.1 | 8 | 805 | |||
XM_017006963.1 | XP_016862452.1 | 8 | 986 | |||
XM_017006964.1 | XP_016862453.1 | 7 | 1856 | |||
GenBank mRNA | AB209871.1 | - | 7 | 1116 | ||
BC140904.1 | - | 7 | 1128 | 9056716 | ||
BC144110.1 | - | 7 | 1128 | 9052630 | ||
L09247.1 | - | 7 | 830 |