Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRS SIRNA Assays ›
Gene Symbol:
PTPRS
Gene Name:
protein tyrosine phosphatase, receptor type S
Gene Aliases:
PTPSIGMA, R-PTP-S, R-PTP-sigma
Chromosome Location:
Chr.19: 5205503 - 5340803 on Build GRCh38
UniGene ID:
Hs.744928
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142411
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002850.3
NP_002841.3
5
781
NM_130853.2
NP_570923.2
5
781
NM_130854.2
NP_570924.2
5
781
NM_130855.2
NP_570925.2
5
781
XM_005259600.2
XP_005259657.1
5
749
XM_005259606.2
XP_005259663.1
5
749
XM_005259607.2
XP_005259664.1
5
749
XM_005259609.1
XP_005259666.1
5
748
XM_005259610.1
XP_005259667.1
5
748
XM_011528157.2
XP_011526459.1
4
619
XM_017027065.1
XP_016882554.1
5
749
XM_017027066.1
XP_016882555.1
5
767
XM_017027067.1
XP_016882556.1
5
665
XM_017027068.1
XP_016882557.1
5
749
XM_017027069.1
XP_016882558.1
5
749
XM_017027070.1
XP_016882559.1
5
749
XM_017027071.1
XP_016882560.1
5
749
XM_017027072.1
XP_016882561.1
5
749
XM_017027074.1
XP_016882563.1
5
748
XM_017027075.1
XP_016882564.1
5
748
XM_017027076.1
XP_016882565.1
5
748
GenBank mRNA
AB209333.1
-
5
781
BC104812.1
-
4
621
8143815
BC143286.1
-
4
621
BC143287.1
-
4
621
9051790
U35234.1
-
4
655
U40317.1
-
4
569
U41725.1
-
4
569
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
chemical synaptic transmission
spinal cord development
cerebellum development
hippocampus development
cerebral cortex development
corpus callosum development
peptidyl-tyrosine dephosphorylation
establishment of endothelial intestinal barrier
transmembrane receptor protein tyrosine phosphatase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-gx66z:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline