Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278639.1 | NP_001265568.1 | 2 | 558 | ||
NM_001278640.1 | NP_001265569.1 | 2 | 306 | |||
NM_001278641.1 | NP_001265570.1 | 2 | 426 | |||
NM_002882.3 | NP_002873.1 | 2 | 306 | |||
XM_011530289.2 | XP_011528591.1 | 2 | 328 | |||
XM_011530290.2 | XP_011528592.1 | 2 | 342 | |||
XM_011530291.2 | XP_011528593.1 | 2 | 620 | |||
XM_017028890.1 | XP_016884379.1 | 2 | 328 | |||
XM_017028891.1 | XP_016884380.1 | 2 | 421 | |||
XM_017028892.1 | XP_016884381.1 | 2 | 212 | |||
GenBank mRNA | AK097659.1 | - | 2 | 523 | ||
AK223506.1 | - | 2 | 225 | |||
AK293502.1 | - | 2 | 173 | |||
AK300927.1 | - | 2 | 225 | |||
BG717186.1 | - | 2 | 175 | |||
BI916757.1 | - | 2 | 426 | |||
CA489474.1 | - | 2 | 248 | |||
CR456556.1 | - | 2 | 134 | |||
D38076.1 | - | 2 | 229 | |||
DB459798.1 | - | 2 | 308 | |||
X83617.1 | - | 2 | 182 |