Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278651.1 | NP_001265580.1 | 7 | 1273 | ||
NM_001317930.1 | NP_001304859.1 | 6 | 727 | |||
NM_002883.3 | NP_002874.1 | 6 | 812 | |||
XM_005261695.1 | XP_005261752.1 | 6 | 940 | |||
XM_005261696.1 | XP_005261753.1 | 6 | 711 | |||
XM_006724289.3 | XP_006724352.1 | 6 | 715 | |||
XM_011530294.2 | XP_011528596.1 | 6 | 2421 | |||
XM_011530295.2 | XP_011528597.1 | 6 | 1120 | |||
XM_011530297.1 | XP_011528599.1 | 6 | 940 | |||
XM_017028893.1 | XP_016884382.1 | 7 | 2051 | |||
XM_017028894.1 | XP_016884383.1 | 7 | 2189 | |||
XM_017028895.1 | XP_016884384.1 | 6 | 2551 | |||
XM_017028896.1 | XP_016884385.1 | 7 | 939 | |||
XM_017028897.1 | XP_016884386.1 | 6 | 2031 | |||
XM_017028898.1 | XP_016884387.1 | 8 | 1964 | |||
GenBank mRNA | AB058738.1 | - | 5 | 1996 | ||
AK303524.1 | - | 6 | 745 | |||
BC014044.2 | - | 6 | 727 | 3506039 | ||
BC041396.2 | - | 7 | 1283 | 5277434 | ||
CR456557.1 | - | 5 | 569 | |||
X82260.1 | - | 5 | 508 |