Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAP1GAP SIRNA Assays ›
Gene Symbol:
RAP1GAP
Gene Name:
RAP1 GTPase activating protein
Gene Aliases:
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location:
Chr.1: 21596215 - 21669444 on Build GRCh38
UniGene ID:
Hs.148178
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142443
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAP1GAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145657.1
NP_001139129.1
8
549
NM_001145658.1
NP_001139130.1
7
409
NM_002885.2
NP_002876.2
7
442
XM_005245955.3
XP_005246012.2
7
454
XM_006710804.2
XP_006710867.2
7
454
XM_006710805.2
XP_006710868.2
7
454
XM_017001965.1
XP_016857454.1
6
456
XM_017001966.1
XP_016857455.1
6
456
XM_017001967.1
XP_016857456.1
7
454
XM_017001968.1
XP_016857457.1
7
489
XM_017001969.1
XP_016857458.1
6
456
XM_017001970.1
XP_016857459.1
6
456
XM_017001971.1
XP_016857460.1
7
408
XM_017001972.1
XP_016857461.1
7
454
XM_017001973.1
XP_016857462.1
6
487
XM_017001974.1
XP_016857463.1
7
454
XM_017001975.1
XP_016857464.1
6
456
XM_017001976.1
XP_016857465.1
7
454
XM_017001977.1
XP_016857466.1
7
454
XM_017001978.1
XP_016857467.1
8
603
XM_017001979.1
XP_016857468.1
9
607
XM_017001980.1
XP_016857469.1
7
408
XM_017001981.1
XP_016857470.1
9
583
XM_017001982.1
XP_016857471.1
8
603
XM_017001983.1
XP_016857472.1
10
815
XM_017001984.1
XP_016857473.1
9
583
XM_017001985.1
XP_016857474.1
7
408
XM_017001986.1
XP_016857475.1
9
620
XM_017001987.1
XP_016857476.1
8
549
XM_017001988.1
XP_016857477.1
8
520
XM_017001989.1
XP_016857478.1
9
620
XM_017001990.1
XP_016857479.1
8
505
XM_017001991.1
XP_016857480.1
8
543
GenBank mRNA
AB003930.1
-
5
383
AB007943.3
-
8
487
BC035030.1
-
4
198
5261027
BC054490.1
-
7
349
5767775
BP200029.1
-
7
409
BP283134.1
-
7
442
CV574545.1
-
7
399
M64788.1
-
7
385
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
regulation of GTPase activity
positive regulation of GTPase activity
negative regulation of neuron differentiation
regulation of small GTPase mediated signal transduction
negative regulation of microvillus assembly
negative regulation of GTP binding
negative regulation of thyroid gland epithelial cell proliferation
cellular response to glial cell derived neurotrophic factor
GTPase activity
GTPase activator activity
protein binding
Ras GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline