Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002887.3 | NP_002878.2 | 3 | 316 | ||
GenBank mRNA | AK222797.1 | - | 3 | 292 | ||
AK302161.1 | - | 3 | 283 | |||
AK314795.1 | - | 3 | 304 | |||
AU125370.1 | - | 3 | 317 | |||
BC000528.2 | - | 3 | 294 | 2961565 | ||
BC014619.2 | - | 3 | 294 | 2961565 | ||
BT007394.1 | - | 3 | 275 | |||
S80343.1 | - | 3 | 302 |