Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002901.2 | NP_002892.1 | 2 | 602 | ||
GenBank mRNA | AK094360.1 | - | 1 | 692 | ||
AK129791.1 | - | 2 | 629 | |||
BC010120.2 | - | 2 | 471 | 3836263 | ||
D42073.1 | - | 2 | 388 | |||
FJ224346.1 | - | 2 | 388 |