Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260492.1 | NP_001247421.1 | 5, 6 | 783 | ||
NM_001260493.1 | NP_001247422.1 | 5, 6 | 783 | |||
NM_001260496.1 | NP_001247425.1 | 4, 5 | 687 | |||
NM_002906.3 | NP_002897.1 | 5, 6 | 783 | |||
GenBank mRNA | AK312903.1 | - | 5, 6 | 685 | ||
BC002626.2 | - | 5, 6 | 667 | |||
BC020751.1 | - | 5, 6 | 683 | |||
BC047109.1 | - | 5, 6 | 683 | 5284438 | ||
DQ916738.1 | - | 5, 6 | 783 | |||
DQ916739.1 | - | 4, 5 | 687 | |||
DQ916741.1 | - | 5, 6 | 783 | |||
DQ916742.1 | - | 5, 6 | 783 | |||
L02320.1 | - | 4, 5 | 503 |