Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004057.2 | NP_004048.1 | 2 | 149 | ||
XM_017029841.1 | XP_016885330.1 | 2 | 616 | |||
GenBank mRNA | BC112174.1 | - | 1 | 115 | 8327642 | |
L13220.1 | - | 2 | 152 | |||
X65869.1 | - | 2 | 149 |