Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS7 SIRNA Assays ›
Gene Symbol:
RGS7
Gene Name:
regulator of G-protein signaling 7
Gene Aliases:
-
Chromosome Location:
Chr.1: 240775193 - 241357374 on Build GRCh38
UniGene ID:
Hs.655739
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142488
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282773.1
NP_001269702.1
14
1540
NM_001282775.1
NP_001269704.1
16
1699
NM_001282778.1
NP_001269707.1
16
1699
NM_002924.5
NP_002915.3
16
1699
XM_005273218.3
XP_005273275.1
16
1843
XM_006711800.3
XP_006711863.1
15
1634
XM_011544246.2
XP_011542548.1
15
1293
XM_011544247.2
XP_011542549.1
15
1338
XM_017002001.1
XP_016857490.1
15
1792
XM_017002002.1
XP_016857491.1
17
1718
XM_017002003.1
XP_016857492.1
15
1792
XM_017002004.1
XP_016857493.1
15
1792
XM_017002005.1
XP_016857494.1
16
2734
XM_017002006.1
XP_016857495.1
17
1764
XM_017002007.1
XP_016857496.1
17
1695
XM_017002008.1
XP_016857497.1
17
1723
XM_017002009.1
XP_016857498.1
15
1623
XM_017002010.1
XP_016857499.1
16
1408
XM_017002011.1
XP_016857500.1
13
2805
XM_017002012.1
XP_016857501.1
13
2803
XM_017002013.1
XP_016857502.1
13
2802
GenBank mRNA
AF090116.1
-
16
1554
AF090117.1
-
9
810
AF493930.1
-
13
1158
AF493931.1
-
15
1317
AK294234.1
-
16
1533
AK294354.1
-
16
1674
AK314862.1
-
16
1560
AY587875.1
-
15
1317
BC022009.1
-
16
1652
4823760
U32439.1
-
14
1171
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
G-protein coupled receptor signaling pathway
negative regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
G-protein beta-subunit binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0