Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNH1 SIRNA Assays ›
Gene Symbol:
RNH1
Gene Name:
ribonuclease/angiogenin inhibitor 1
Gene Aliases:
RAI, RNH
Chromosome Location:
Chr.11: 494512 - 507283 on Build GRCh38
UniGene ID:
Hs.530687
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142494
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002939.3
NP_002930.2
11
1947
NM_203383.1
NP_976317.1
11
1879
NM_203384.1
NP_976318.1
10
1706
NM_203385.1
NP_976319.1
10
1774
NM_203386.2
NP_976320.1
11
1837
NM_203387.2
NP_976321.1
11
1905
NM_203388.2
NP_976322.1
10
1664
NM_203389.2
NP_976323.1
10
1732
XM_011520255.1
XP_011518557.1
11
1882
XM_011520257.2
XP_011518559.1
11
2130
XM_011520258.2
XP_011518560.1
11
2098
XM_011520259.2
XP_011518561.1
10
1647
XM_011520260.2
XP_011518562.1
11
1806
XM_011520261.2
XP_011518563.1
11
1880
XM_011520262.2
XP_011518564.1
11
2063
XM_011520263.1
XP_011518565.1
10
1614
XM_017018106.1
XP_016873595.1
11
1947
GenBank mRNA
AB209196.1
-
9
3197
AK057144.1
-
2
1996
AK311379.1
-
7
996
AL161967.1
-
9
2882
BC000677.2
-
10
1634
3349381
BC003506.1
-
10
1647
2959514
BC011186.1
-
9
1432
BC011500.1
-
10
1585
4155804
BC014629.2
-
10
1634
3029730
BC024037.1
-
10
1647
2959514
BC047730.1
-
11
1926
5788220
BQ650217.1
-
3
456
M22414.1
-
10
1595
M36717.1
-
11
1834
X13973.1
-
11
1834
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
negative regulation of catalytic activity
regulation of angiogenesis
protein binding
ribonuclease inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline