Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005861.2 | NP_001005861.1 | 8, 9 | 1122 | ||
NM_002958.3 | NP_002949.2 | 8, 9 | 1113 | |||
XM_017007016.1 | XP_016862505.1 | 3, 4 | 619 | |||
GenBank mRNA | BC021700.2 | - | 3, 4 | 388 | ||
S59184.1 | - | 8, 9 | 1146 | |||
X69970.1 | - | 8, 9 | 1113 | |||
X96588.1 | - | 9, 10 | 1018 |