Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144663.1 | NP_001138135.1 | 18 | 3201 | ||
NM_004063.3 | NP_004054.3 | 18 | 3200 | |||
XM_011516790.1 | XP_011515092.1 | 17 | 2959 | |||
GenBank mRNA | AK130441.1 | - | 2 | 802 | ||
U07969.1 | - | 18 | 3162 | |||
X83228.1 | - | 18 | 3167 |