Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002972.3 | NP_002963.2 | 5 | 737 | ||
XM_005261935.2 | XP_005261992.1 | 5 | 685 | |||
XM_011530709.2 | XP_011529011.1 | 5 | 555 | |||
XM_011530710.2 | XP_011529012.1 | 5 | 551 | |||
XM_017028905.1 | XP_016884394.1 | 5 | 554 | |||
GenBank mRNA | AB209682.1 | - | 4 | 425 | ||
AK057985.1 | - | 6 | 562 | |||
BC040031.1 | - | 5 | 688 | |||
BC087612.1 | - | 5 | 676 | 6671883 | ||
BI821356.1 | - | 5 | 696 | |||
CR749672.1 | - | 3 | 353 |