Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002972.3 | NP_002963.2 | 30 | 4287 | ||
XM_005261935.2 | XP_005261992.1 | 29 | 4157 | |||
XM_011530709.2 | XP_011529011.1 | 30 | 4105 | |||
XM_011530710.2 | XP_011529012.1 | 30 | 4101 | |||
XM_017028905.1 | XP_016884394.1 | 29 | 4026 | |||
GenBank mRNA | BC009268.1 | - | 8 | 1168 | ||
BC087612.1 | - | 30 | 4226 | 6671883 | ||
BG766677.1 | - | 5 | 644 | |||
U93181.1 | - | 23 | 3284 |