Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CX3CL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002996.4 | NP_002987.1 | 2 | 197 | ||
GenBank mRNA | AB209037.1 | - | 1 | 2679 | ||
AK222692.1 | - | 2 | 187 | |||
AK223351.1 | - | 2 | 168 | |||
AK313806.1 | - | 2 | 168 | |||
BC001163.1 | - | 2 | 156 | 3503870 | ||
BC016164.1 | - | 2 | 156 | 3502121 | ||
CR457426.1 | - | 2 | 89 | |||
HY114023.1 | - | 2 | 198 | |||
U84487.1 | - | 2 | 168 | |||
U91835.1 | - | 2 | 168 |