Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003005.3 | NP_002996.2 | 16 | 2515 | ||
XM_005245435.1 | XP_005245492.1 | 16 | 2512 | |||
XM_005245436.3 | XP_005245493.1 | 16 | 2512 | |||
XM_005245438.1 | XP_005245495.1 | 15 | 2392 | |||
XM_005245439.1 | XP_005245496.1 | 15 | 2392 | |||
XM_005245440.1 | XP_005245497.1 | 15 | 2326 | |||
GenBank mRNA | AK312884.1 | - | 16 | 2497 | ||
BC028067.2 | - | 14 | 2327 | |||
BC068533.1 | - | 16 | 2498 | 30349489 | ||
KU178327.1 | - | 15 | 2447 | |||
KU178328.1 | - | 6 | 724 | |||
M25322.1 | - | 16 | 2488 |