Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELPLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206609.1 | NP_001193538.1 | 2 | 1131 | ||
NM_003006.4 | NP_002997.2 | 2 | 1066 | |||
GenBank mRNA | AK098315.1 | - | 4 | 985 | ||
AK127638.1 | - | 3 | 1017 | |||
AK225644.1 | - | 2 | 962 | |||
AK290395.1 | - | 3 | 1003 | |||
AK295238.1 | - | 3 | 848 | |||
AK298738.1 | - | 2 | 1064 | |||
AK298742.1 | - | 2 | 1103 | |||
AK300057.1 | - | 3 | 958 | |||
BC029782.1 | - | 3 | 1015 | 5180824 | ||
U02297.1 | - | 3 | 935 |