Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144757.1 | NP_001138229.1 | 3 | 440 | ||
NM_003020.3 | NP_003011.1 | 3 | 440 | |||
GenBank mRNA | AK314965.1 | - | 3 | 433 | ||
BC005349.1 | - | 3 | 379 | 3949961 | ||
BC093053.1 | - | 3 | 399 | 30720075 | ||
BP377248.1 | - | 3 | 440 | |||
CR541824.1 | - | 2 | 323 | |||
Y00757.1 | - | 3 | 351 |