Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003021.3 | NP_003012.1 | 7 | 731 | ||
XM_011528178.2 | XP_011526480.1 | 7 | 785 | |||
GenBank mRNA | AF368279.1 | - | 6 | 605 | ||
AF408399.1 | - | 6 | 584 | |||
AJ133129.1 | - | 7 | 626 | |||
AJ223828.1 | - | 6 | 625 | |||
AK293535.1 | - | 7 | 702 | |||
AL050156.1 | - | 7 | 693 | |||
BC000390.2 | - | 7 | 634 | 2821268 | ||
BC002989.1 | - | 7 | 634 | 2821268 | ||
BC005165.2 | - | 7 | 628 | 3532317 | ||
BC008885.2 | - | 7 | 628 | 4302652 | ||
CR533517.1 | - | 6 | 584 | |||
CR542282.1 | - | 6 | 584 |