Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003022.2 | NP_003013.1 | 4 | 1323 | ||
XM_011531013.1 | XP_011529315.1 | 5 | 1127 | |||
XM_011531014.1 | XP_011529316.1 | 6 | 1446 | |||
GenBank mRNA | AF042081.1 | - | 4 | 1002 | ||
AK024892.1 | - | 5 | 1097 | |||
AK129610.1 | - | 4 | 1008 | |||
AL136718.1 | - | 4 | 1024 | |||
BC016709.1 | - | 4 | 1029 | 4105289 | ||
BC103762.1 | - | 4 | 1148 | 6179172 | ||
CN272007.1 | - | 1 | 220 | |||
FJ224348.1 | - | 4 | 1002 |