Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122681.1 | NP_001116153.1 | 12 | 1634 | ||
NM_001145855.1 | NP_001139327.1 | 12 | 1777 | |||
NM_001145856.1 | NP_001139328.1 | 12 | 1724 | |||
NM_003023.4 | NP_003014.3 | 12 | 1775 | |||
GenBank mRNA | AB000462.1 | - | 12 | 1775 | ||
AB000463.1 | - | 13 | 1954 | |||
AF000936.1 | - | 11 | 1514 | |||
AK124073.1 | - | 7 | 1620 | |||
AK126537.1 | - | 6 | 2343 | |||
AK225497.1 | - | 12 | 1636 | |||
AK299996.1 | - | 12 | 1693 | |||
AK312286.1 | - | 12 | 1572 | |||
BC022996.1 | - | 12 | 1630 | 5103986 | ||
U56386.1 | - | 12 | 1722 |