Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003061.2 | NP_003052.2 | 3 | 564 | ||
GenBank mRNA | AB011537.3 | - | 3 | 564 | ||
AB017167.1 | - | 3 | 544 | |||
AY029183.1 | - | 3 | 261 | |||
BC146761.1 | - | 3 | 564 | 9007200 | ||
BC146851.1 | - | 3 | 477 | 9056779 |
Gene Symbol: ARHGAP19-SLIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037909.1 | 13 | 1635 | |||
GenBank mRNA | AY336750.1 | - | 13 | 1600 |