Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135058.1 | NP_001128530.1 | 11 | 2309 | ||
NM_004086.2 | NP_004077.1 | 12 | 1985 | |||
XM_017021071.1 | XP_016876560.1 | 12 | 2219 | |||
XM_017021072.1 | XP_016876561.1 | 11 | 2358 | |||
XM_017021073.1 | XP_016876562.1 | 4 | 1367 | |||
GenBank mRNA | AF006740.1 | - | 12 | 1961 | ||
AK123362.1 | - | 10 | 2114 | |||
AK130273.1 | - | 6 | 1566 | |||
AY358900.1 | - | 11 | 2300 | |||
BC018827.2 | - | 12 | 1947 |