Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003063.2 | NP_003054.1 | 2 | 490 | ||
GenBank mRNA | BC005261.1 | - | 2 | 485 | 3934241 | |
BC094685.1 | - | 2 | 472 | 6203008 | ||
BP233121.1 | - | 2 | 490 | |||
F30315.1 | - | 1 | 104 | |||
U96094.1 | - | 2 | 484 |