Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003086.2 | NP_003077.2 | 12 | 1283 | ||
XM_005266096.2 | XP_005266153.1 | 13 | 1374 | |||
XM_006717241.2 | XP_006717304.1 | 13 | 1358 | |||
XM_006717242.3 | XP_006717305.1 | 13 | 2287 | |||
XM_017015058.1 | XP_016870547.1 | 12 | 2803 | |||
GenBank mRNA | AF032387.1 | - | 12 | 1649 |