Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003098.2 | NP_003089.1 | 4 | 1076 | ||
XM_005260517.1 | XP_005260574.1 | 4 | 1037 | |||
XM_011529007.1 | XP_011527309.1 | 4 | 1037 | |||
XM_011529008.1 | XP_011527310.1 | 4 | 1037 | |||
GenBank mRNA | AK095942.1 | - | 4 | 636 | ||
AK291994.1 | - | 4 | 947 | |||
BC026215.2 | - | 4 | 916 | 4413197 | ||
BC113813.1 | - | 2 | 224 | |||
S81737.1 | - | 4 | 936 | |||
U40571.1 | - | 4 | 841 |