Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4EBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004095.3 | NP_004086.1 | 3 | 509 | ||
GenBank mRNA | AB044548.1 | - | 3 | 482 | ||
BC004459.1 | - | 3 | 501 | 2820216 | ||
BC058073.1 | - | 3 | 449 | 5190933 |