Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130438.2 | NP_001123910.1 | 18 | 2632 | ||
NM_001195532.1 | NP_001182461.1 | 18 | 2632 | |||
NM_003127.3 | NP_003118.2 | 18 | 2632 | |||
XM_006717245.1 | XP_006717308.1 | 18 | 2539 | |||
XM_006717246.1 | XP_006717309.1 | 18 | 2539 | |||
XM_006717247.2 | XP_006717310.1 | 18 | 2539 | |||
XM_006717248.2 | XP_006717311.1 | 18 | 2538 | |||
XM_006717249.1 | XP_006717312.1 | 18 | 2539 | |||
XM_006717250.1 | XP_006717313.1 | 18 | 2539 | |||
XM_006717251.2 | XP_006717314.1 | 18 | 2626 | |||
XM_006717252.2 | XP_006717315.1 | 18 | 2539 | |||
XM_006717253.1 | XP_006717316.1 | 18 | 2539 | |||
XM_006717254.2 | XP_006717317.1 | 18 | 2626 | |||
XM_017015059.1 | XP_016870548.1 | 18 | 2617 | |||
XM_017015060.1 | XP_016870549.1 | 18 | 2539 | |||
XM_017015061.1 | XP_016870550.1 | 18 | 2597 | |||
GenBank mRNA | AB191262.1 | - | 18 | 2597 | ||
AB209860.1 | - | 18 | 2594 | |||
AK294753.1 | - | 19 | 2656 | |||
AK300386.1 | - | 18 | 2600 | |||
AK307676.1 | - | 18 | 2600 | |||
BC053521.1 | - | 18 | 2581 | 5529391 | ||
J05243.1 | - | 18 | 2592 | |||
M18627.1 | - | 3 | 466 | |||
M24773.1 | - | 3 | 466 | |||
U26396.1 | - | 1 | 62 | |||
U83867.1 | - | 18 | 2559 |