Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278656.1 | NP_001265585.1 | 4 | 1211 | ||
NM_004175.4 | NP_004166.1 | 4 | 1225 | |||
NR_103819.1 | 3 | 1032 | ||||
GenBank mRNA | BC034447.1 | - | 4 | 908 |