Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318798.1 | NP_001305727.1 | 7, 8 | 711 | ||
NM_001318800.1 | NP_001305729.1 | 7, 8 | 995 | |||
NM_004186.4 | NP_004177.3 | 8, 9 | 972 | |||
XM_005265381.4 | XP_005265438.1 | 8, 9 | 1102 | |||
XM_005265382.4 | XP_005265439.1 | 7, 8 | 812 | |||
XM_006713289.3 | XP_006713352.1 | 2, 3 | 584 | |||
XM_006713290.3 | XP_006713353.1 | 8, 9 | 905 | |||
XM_011533998.2 | XP_011532300.1 | 8, 9 | 904 | |||
XM_011534000.2 | XP_011532302.1 | 8, 9 | 905 | |||
GenBank mRNA | AB209259.1 | - | 7, 8 | 665 | ||
BC042914.1 | - | 8, 9 | 968 | 5477898 | ||
U33920.1 | - | 7, 8 | 848 | |||
U38276.1 | - | 7, 8 | 992 |