Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258012.3 | NP_001244941.2 | 12 | 1877 | ||
NM_001258013.3 | NP_001244942.2 | 13 | 1841 | |||
NM_001258014.3 | NP_001244943.2 | 12 | 1724 | |||
NM_001258015.2 | NP_001244944.1 | 11 | 1628 | |||
NM_001258016.3 | NP_001244945.2 | 11 | 1693 | |||
NM_004110.5 | NP_004101.3 | 12 | 1766 | |||
NM_024417.4 | NP_077728.3 | 12 | 1748 | |||
NR_047576.2 | 12 | 1848 | ||||
XM_006721772.1 | XP_006721835.1 | 12 | 1696 | |||
XM_011524527.1 | XP_011522829.1 | 12 | 1714 | |||
XM_011524528.1 | XP_011522830.1 | 12 | 1803 | |||
XM_011524529.1 | XP_011522831.1 | 12 | 1806 | |||
XM_011524531.2 | XP_011522833.1 | 11 | 1699 | |||
XM_011524532.2 | XP_011522834.1 | 11 | 1706 | |||
XM_011524533.2 | XP_011522835.1 | 11 | 1674 | |||
GenBank mRNA | AK094120.1 | - | 12 | 2026 | ||
AK098648.1 | - | 12 | 1685 | |||
AK293208.1 | - | 13 | 1786 | |||
AK293210.1 | - | 12 | 1785 | |||
AK295307.1 | - | 12 | 1877 | |||
AK298908.1 | - | 11 | 1571 | |||
AK301779.1 | - | 12 | 1661 | |||
AK301977.1 | - | 11 | 1693 | |||
BC002960.1 | - | 12 | 1687 | 3537008 | ||
BC018911.2 | - | 12 | 1694 | |||
BC034763.1 | - | 12 | 2349 | |||
BC063493.1 | - | 12 | 1686 | 5404814 | ||
J03826.1 | - | 12 | 1699 |