Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005399.4 | NP_005390.1 | 3, 4 | 515 | ||
XM_011509729.1 | XP_011508031.1 | 3, 4 | 515 | |||
XM_017001697.1 | XP_016857186.1 | 3, 4 | 515 | |||
GenBank mRNA | AJ224538.1 | - | 2, 3 | 331 | ||
AK292820.1 | - | 3, 4 | 396 | |||
AK310982.1 | - | 3, 4 | 396 | |||
BC053610.1 | - | 3, 4 | 470 | 6091476 | ||
KJ534919.1 | - | 2, 3 | 331 | |||
KJ535025.1 | - | 2, 3 | 331 |