Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005399.4 | NP_005390.1 | 8 | 1796 | ||
NR_103870.1 | 6 | 1508 | ||||
NR_103871.1 | 7 | 1629 | ||||
XM_011509729.1 | XP_011508031.1 | 9 | 1833 | |||
XM_017001697.1 | XP_016857186.1 | 7 | 1675 | |||
GenBank mRNA | AK023717.1 | - | 1 | 411 | ||
BC053610.1 | - | 8 | 1751 | 6091476 |