Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102445.2 | NP_001095915.1 | 6 | 2257 | ||
NM_001113380.1 | NP_001106851.1 | 5 | 1832 | |||
NM_001113381.1 | NP_001106852.1 | 4 | 1701 | |||
NM_005613.5 | NP_005604.1 | 5 | 2148 | |||
GenBank mRNA | AB209019.1 | - | 1 | 4220 | ||
AK093959.1 | - | 4 | 1588 | |||
AK096204.1 | - | 3 | 2758 | |||
BC000737.1 | - | 5 | 1718 | 3510260 | ||
BC051869.1 | - | 5 | 1730 | 5786695 | ||
DQ346662.1 | - | 5 | 1757 | |||
DQ346663.1 | - | 5 | 1972 | |||
DQ346664.1 | - | 6 | 2084 |