Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005619.4 | NP_005610.1 | 11 | 2037 | ||
NM_206900.2 | NP_996783.1 | 10 | 1818 | |||
NM_206901.2 | NP_996784.1 | 7 | 860 | |||
GenBank mRNA | AF004222.1 | - | 11 | 1936 | ||
AF004223.1 | - | 8 | 1086 | |||
AF004224.1 | - | 7 | 828 | |||
AF038540.1 | - | 8 | 1267 | |||
BC073874.1 | - | 11 | 1979 | 5242827 |