Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 7, 8 | 1308 | ||
NM_001143677.1 | NP_001137149.1 | 5, 6 | 559 | |||
NM_001143678.1 | NP_001137150.1 | 5, 6 | 738 | |||
NM_005627.3 | NP_005618.2 | 5, 6 | 514 | |||
XM_011536071.1 | XP_011534373.1 | 8, 9 | 1090 | |||
GenBank mRNA | AF153609.1 | - | 5, 6 | 467 | ||
AK055077.1 | - | 7, 8 | 1308 | |||
AK098509.1 | - | 5, 6 | 482 | |||
AK293383.1 | - | 5, 6 | 548 | |||
AK298688.1 | - | 5, 6 | 797 | |||
BC001263.1 | - | 5, 6 | 467 | 3459056 | ||
EU518415.1 | - | 7, 8 | 710 | |||
FM205707.1 | - | 5, 6 | 502 | |||
FM205708.1 | - | 5, 6 | 537 | |||
FM205709.1 | - | 7, 8 | 1157 | |||
FM205710.1 | - | 4, 5 | 626 | |||
Y10032.1 | - | 5, 6 | 467 |