Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 12 | 1798 | ||
NM_001143677.1 | NP_001137149.1 | 10 | 1049 | |||
NM_001143678.1 | NP_001137150.1 | 10 | 1228 | |||
NM_001291995.1 | NP_001278924.1 | 9 | 872 | |||
NM_005627.3 | NP_005618.2 | 10 | 1004 | |||
XM_011536071.1 | XP_011534373.1 | 13 | 1580 | |||
GenBank mRNA | AF153609.1 | - | 10 | 957 | ||
AK055077.1 | - | 12 | 1798 | |||
AK098509.1 | - | 10 | 972 | |||
AK293383.1 | - | 10 | 1038 | |||
AK295422.1 | - | 9 | 840 | |||
AK298688.1 | - | 10 | 1287 | |||
BC001263.1 | - | 10 | 957 | 3459056 | ||
BX649005.1 | - | 2 | 3577 | |||
EU518415.1 | - | 12 | 1200 | |||
FM205707.1 | - | 10 | 992 | |||
FM205708.1 | - | 10 | 1027 | |||
FM205709.1 | - | 12 | 1647 | |||
FM205710.1 | - | 9 | 1116 | |||
Y10032.1 | - | 10 | 957 |