Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007559.2 | NP_001007560.1 | 11 | 2235 | ||
NM_001308201.1 | NP_001295130.1 | 11 | 2210 | |||
NM_005637.3 | NP_005628.2 | 10 | 2142 | |||
XM_006722527.2 | XP_006722590.1 | 12 | 2261 | |||
XM_011526145.1 | XP_011524447.1 | 11 | 2203 | |||
XM_011526147.2 | XP_011524449.1 | 13 | 2486 | |||
XM_011526148.2 | XP_011524450.1 | 13 | 2473 | |||
XM_011526149.2 | XP_011524451.1 | 13 | 2498 | |||
XM_011526150.2 | XP_011524452.1 | 12 | 2358 | |||
XM_011526151.2 | XP_011524453.1 | 13 | 2487 | |||
XM_011526152.2 | XP_011524454.1 | 11 | 2265 | |||
GenBank mRNA | AK296949.1 | - | 11 | 2193 | ||
X79201.1 | - | 10 | 2079 |