Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004135.3 | NP_004126.1 | 7 | 682 | ||
NM_174869.2 | NP_777358.1 | 7 | 682 | |||
GenBank mRNA | AB209206.1 | - | 2 | 3293 | ||
BC000933.2 | - | 7 | 528 | 3447003 | ||
BC001902.1 | - | 7 | 523 | 3543839 | ||
BI831096.1 | - | 7 | 680 | |||
U40272.1 | - | 7 | 677 | |||
U69268.1 | - | 3 | 233 | |||
Z68907.1 | - | 7 | 564 |